missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCK Rabbit anti-Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15629-1-AP-20UL
This item is not returnable.
View return policy
Description
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Alternative splicing of this gene results in three tissue-specific forms of glucokinase, one found in pancreatic islet beta cells and two found in liver. The protein localizes to the outer membrane of mitochondria. In contrast to other forms of hexokinase, this enzyme is not inhibited by its product glucose-6-phosphate but remains active while glucose is abundant. Mutations in this gene have been associated with non-insulin dependent diabetes mellitus (NIDDM), maturity onset diabetes of the young, type 2 (MODY2) and persistent hyperinsulinemic hypoglycemia of infancy (PHHI).Specifications
| GCK | |
| Polyclonal | |
| Unconjugated | |
| Gck | |
| GCK, GK, GLK, Glucokinase, glucokinase (hexokinase 4), Hexokinase 4, Hexokinase D, Hexokinase type IV, HHF3, HK IV, HK4, HKIV, HXKP, MODY2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103988, 24385 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17712, P52792 | |
| Gck | |
| GCK Fusion Protein Ag7904 | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction