missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX GCK Protein, Active

Product Code. 17390644
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17390644

Brand: Eurofins DiscoverX 14743M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal GST- tagged, recombinant, human GCK amino acids 1-473, expressed by baculovirus in Sf21 cells, for use in Kinase Assays.

GCK (Glucokinase) is a member of the hexokinase family of proteins. Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. In contrast to other forms of hexokinase, this enzyme is not inhibited by its product glucose-6-phosphate but remains active while glucose is abundant. The use of multiple promoters and alternative splicing of this gene result in distinct protein isoforms that exhibit tissue-specific expression in the pancreas and liver. In the pancreas, this enzyme plays a role in glucose-stimulated insulin secretion, while in the liver, this enzyme is important in glucose uptake and conversion to glycogen. Mutations in this gene that alter enzyme activity have been associated with multiple types of diabetes and hyperinsulinemic hypoglycemia.

GenBank BC047865

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 80.6 kDa
Quantity 250 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥38%
Protein GCK, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.