Learn More
Invitrogen™ GCK Monoclonal Antibody (4G6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515878
Description
MA5-15878 targets GCK in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15878 immunogen is purified recombinant fragment of human GCK expressed in E. Coli. . MA5-15878 detects GCK which has a predicted molecular weight of approximately 52kDa.
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Alternative splicing of this gene results in three tissue-specific forms of glucokinase, one found in pancreatic islet beta cells and two found in liver. The protein localizes to the outer membrane of mitochondria. In contrast to other forms of hexokinase, this enzyme is not inhibited by its product glucose-6-phosphate but remains active while glucose is abundant. Mutations in this gene have been associated with non-insulin dependent diabetes mellitus (NIDDM), maturity onset diabetes of the young, type 2 (MODY2) and persistent hyperinsulinemic hypoglycemia of infancy (PHHI).
Specifications
| GCK | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P35557 | |
| GCK | |
| Purified recombinant fragment of human GCK expressed in E. Coli. | |
| RUO | |
| 2645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 4G6 | |
| Unconjugated | |
| GCK | |
| ATP:D-hexose 6-phosphotransferase; FGQTL3; GC kinase; Gck; Gk; GLK; Gls006; glucokinase; glucokinase (hexokinase 4); glucokinase variant 3; GLUKA; hexokinase 4; hexokinase D, pancreatic isozyme; hexokinase type IV; Hexokinase-4; hexokinase-D; HHF3; HK IV; HK4; HKIV; Hlb62; HXKP; LGLK; MEKKK 2; MODY2; RNGK2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.