missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCHFR Monoclonal Antibody (846011)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561884

Product Code. 30534430

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

Mediates tetrahydrobiopterin inhibition of GTP cyclohydrolase 1. This inhibition is reversed by L-phenylalanine.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCHFR
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P30047
GCHFR
E. coli-derived recombinant human GCHFR, Met1-Glu84.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
846011
Unconjugated
GCHFR
2010323F13Rik; GCHFR; GFRP; GTP cyclohydrolase 1 feedback regulatory protein; GTP cyclohydrolase I feedback regulator; GTP cyclohydrolase I feedback regulatory protein; HsT16933; p35
Mouse
Protein A/G
RUO
2644
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.