missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GCHFR Antibody (GCHFR/7734), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26792-20ug

 View more versions of this product

Product Code. 30043359

  • £178.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GCHFR Monoclonal antibody specifically detects GCHFR in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCHFR
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
GFRPP35, GTP cyclohydrolase I feedback regulator, GTP cyclohydrolase I feedback regulatory proteinGTP cyclohydrolase 1 feedback regulatory protein, HsT16933, MGC138467, MGC138469, p35
Recombinant fragment human GCHFR protein (exact sequence is proprietary)
20 μg
metabolism
2644
Store at 4°C.
IgG2 κ
Western Blot, Immunohistochemistry (Paraffin)
GCHFR/7734
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.