missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103865
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GCH1.
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with malignant hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
Specifications
| GCH1 | |
| Polyclonal | |
| Unconjugated | |
| GCH1 | |
| dystonia 14; DYT14; DYT5; DYT5a; GCH; Gch1; GTP cyclohydrolase 1; GTP cyclohydrolase I; GTPCH; GTP-CH; GTPCH1; GTP-CH-1; GTP-CH-I; guanosine 5'-triphosphate cyclohydrolase I; HPABH4B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14528, 2643, 29244 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22288, P30793, Q05915 | |
| GCH1 | |
| A synthesized peptide derived from human GCH1(Accession P30793), corresponding to amino acid residues A36-P86. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction