missing translation for 'onlineSavingsMsg'
Learn More

GCH1 Antibody (1F5), Novus Biologicals™

Product Code. 18330649 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330649 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330649 Supplier Novus Biologicals Supplier No. H00002643M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GCH1 Monoclonal antibody specifically detects GCH1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCH1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1F5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH25415
Gene Alias dystonia 14, DYT14, DYT5a, DYT5EC 3.5.4.16, GCH, GTP cyclohydrolase 1, GTP cyclohydrolase I, GTPCH1, GTP-CH-1, GTP-CH-I, guanosine 5'-triphosphate cyclohydrolase I, HPABH4B
Host Species Mouse
Immunogen GCH1 (AAH25415.1, 1 a.a. ~ 250 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEKGPVRAPAEKPRGARCSNGFPERDPPRPGPSRPAEKPPRPEAKSAQPADGWKGERPRSEEDNELNLPNLAAAYSSILSSLGENPQRQGLLKTPWRAASAMQFFTKGYQETISDVLNDAIFDEDHDEMVIVKDIDMFSMCEHHLVPFVGKVHIGYLPNKQVLGLSKLARIVEIYSRRLQVQERLTKQIAVAITEALRPAGVGVVVEATHMCMVMRGVQKMNSKTVTSTMLGVFREDPKTREEFLTLIRS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2643
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.