missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCFC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05984
This item is not returnable.
View return policy
Description
GCFC2 Polyclonal specifically detects GCFC2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| GCFC2 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| C2orf3, chromosome 2 open reading frame 3, DNABF, GC bindng factor, GCF, GCFGC binding factor, GC-rich sequence DNA-binding factor, GC-rich sequence DNA-binding factor 2, TCF9, TCF-9, Transcription factor 9, transcription factor 9 (binds GC-rich sequences) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6936 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| NP_003194.3 | |
| GCFC2 | |
| The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human GC-rich sequence DNA-binding factor using the numbering given in entry NP_003194.3 (GeneID 6936). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction