missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCFC2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05985R
This item is not returnable.
View return policy
Description
GCFC2 Polyclonal antibody specifically detects GCFC2 in Human samples. It is validated for Western Blot
Specifications
| GCFC2 | |
| Polyclonal | |
| Western Blot | |
| C2orf3, chromosome 2 open reading frame 3, DNABF, GC bindng factor, GCF, GCFGC binding factor, GC-rich sequence DNA-binding factor, GC-rich sequence DNA-binding factor 2, TCF9, TCF-9, Transcription factor 9, transcription factor 9 (binds GC-rich sequences) | |
| The immunogen recognized by this antibody maps to a region between residue 731 and 781 of human GC-rich sequence DNA-binding factor using the numbering given in entry NP_003194.3 (GeneID 6936). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6936 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction