missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCET2 Mouse anti-Human, Clone: HGAL/2373, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | GCET2 |
|---|---|
| Clone | HGAL/2373 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364704
|
Novus Biologicals
NBP3-13920-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394705
|
Novus Biologicals
NBP3-13920-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCET2 Monoclonal antibody specifically detects GCET2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
| GCET2 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| GAL, GCAT2, germinal center B-cell-expressed transcript 2 protein, germinal center expressed transcript 2, Germinal center-associated lymphoma protein, HGAL, MGC40441 | |
| Recombinant fragment (178 Amino acid residues from around aa1-200) of human GCET2 protein (Uniprot: Q8N6F7) | |
| Primary | |
| Store at 4°C. |
| HGAL/2373 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q8N6F7 | |
| 257144 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title