missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCET2 Antibody (HGAL/2834), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24293AF647
439.45 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
GCET2 Monoclonal antibody specifically detects GCET2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GCET2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| GAL, GCAT2, germinal center B-cell-expressed transcript 2 protein, germinal center expressed transcript 2, Germinal center-associated lymphoma protein, HGAL, MGC40441 | |
| Recombinant fragment (around aa1-200) of human GCET2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Immunology | |
| 257144 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| HGAL/2834 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction