missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GCET2 Monoclonal antibody specifically detects GCET2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | GCET2 |
| Applications | Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | HGAL/2373 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | GAL, GCAT2, germinal center B-cell-expressed transcript 2 protein, germinal center expressed transcript 2, Germinal center-associated lymphoma protein, HGAL, MGC40441 |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (178 Amino acid residues from around aa1-200) of human GCET2 protein (Uniprot: Q8N6F7) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?