missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCDH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542384
This item is not returnable.
View return policy
Description
Peptide sequence: IARQARDMLG GNGISDEYHV IRHAMNLEAV NTYEGTHDIH ALILGRAITG Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family. It catalyzes the oxidative decarboxylation of glutaryl-CoA to crotonyl-CoA and CO(2) in the degradative pathway of L-lysine, L-hydroxylysine, and L-tryptophan metabolism. It uses electron transfer flavoprotein as its electron acceptor. The enzyme exists in the mitochondrial matrix as a homotetramer of 45-kD subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| GCDH | |
| Polyclonal | |
| Unconjugated | |
| GCDH | |
| 9030411L18; ACAD5; AI266902; D17825; GCD; GCDH; glutaryl-CoA dehydrogenase; glutaryl-CoA dehydrogenase, mitochondrial; glutaryl-Coenzyme A dehydrogenase; MGC137369 protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2639 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92947 | |
| GCDH | |
| Synthetic peptide directed towards the C-terminal of human GCDH (aa 381-430). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction