missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54737
This item is not returnable.
View return policy
Description
GCDH Polyclonal specifically detects GCDH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GCDH | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ACAD5, EC 1.3.99, EC 1.3.99.7, GCD, glutaryl-CoA dehydrogenase, glutaryl-CoA dehydrogenase, mitochondrial, glutaryl-Coenzyme A dehydrogenase | |
| Rabbit | |
| 48 kDa | |
| 100 μL | |
| Stem Cell Markers | |
| 2639 | |
| Human, Mouse, Rat, Pig, Canine, Equine, Rabbit, Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q92947 | |
| GCDH | |
| Synthetic peptides corresponding to GCDH(glutaryl-Coenzyme A dehydrogenase) The peptide sequence was selected from the C terminal of GCDH. Peptide sequence IARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITG. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction