missing translation for 'onlineSavingsMsg'
Learn More

GCC185 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18343556 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343556 100 μL 100µL
18361836 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18343556 Supplier Novus Biologicals Supplier No. NBP303436100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GCC185 Polyclonal specifically detects GCC185 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCC185
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 185 kDa Golgi coiled-coil protein, CLL-associated antigen KW-11, CTCL tumor antigen se1-1, GCC protein, 185-kD, GCC185GRIP and coiled-coil domain-containing 2, Golgi coiled-coil protein GCC185, GRIP and coiled-coil domain containing 2, GRIP and coiled-coil domain-containing protein 2, KIAA0336REN53, Ran-binding protein 2-like 4, RANBP2L4, Renal carcinoma antigen NY-REN-53
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GCC185 (NP_852118.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9648
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.