Learn More
Invitrogen™ GCAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140113
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RGAGTWKSER VITSRQGPHI RVDGVSGGIL NFCANNYLGL SSHPEVIQAG LQALEEFGAG LSSVRFICGT Q Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 93%.
The degradation of L-threonine to glycine consists of a two-step biochemical pathway involving the enzymes L-threonine dehydrogenase and 2-amino-3-ketobutyrate coenzyme A ligase. L-Threonine is first converted into 2-amino-3-ketobutyrate by L-threonine dehydrogenase. GCAT is the second enzyme in this pathway, which then catalyzes the reaction between 2-amino-3-ketobutyrate and coenzyme A to form glycine and acetyl-CoA. The enzyme is considered a class II pyridoxal-phosphate-dependent aminotransferase.
Specifications
| GCAT | |
| Polyclonal | |
| Unconjugated | |
| Gcat | |
| 2-amino-3-ketobutyrate coenzyme A ligase; 2-amino-3-ketobutyrate coenzyme A ligase, mitochondrial; 2-amino-3-ketobutyrate-CoA ligase; AI526977; AKB ligase; aminoacetone synthase; aminoacetone synthetase; aminotransferase; glycine acetyltransferase; aminoacetone synthetase; KBL; GCAT; Glycine acetyltransferase; glycine C-acetyltransferase; glycine C-acetyltransferase (2-amino-3-ketobutyrate coenzyme A ligase); glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase); KBL; Unknown (protein for MGC:139551) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23464 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75600 | |
| Gcat | |
| Recombinant protein corresponding to Human GCAT. Recombinant protein control fragment (Product #RP-109052). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.