missing translation for 'onlineSavingsMsg'
Learn More

GCAP2 Antibody (5E7), Novus Biologicals™

Product Code. 18388027 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388027 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388027 Supplier Novus Biologicals Supplier No. H00002979M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GCAP2 Monoclonal antibody specifically detects GCAP2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCAP2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 5E7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002089
Gene Alias DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48
Host Species Mouse
Immunogen GUCA1B (NP_002089, 93 a.a. ~ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KLKWTFKIYDKDGNGCIDRLELLNIVEGIYQLKKACRRELQTEQDQLLTPEEVVDRIFLLVDENGDGQLSLNEFVEGARRDKWVMKMLQMDMNPSSWLAQQRRKSAMF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2979
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.