missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12944-1-AP-20UL
This item is not returnable.
View return policy
Description
Guanylate cyclase-activating protein is a l Ca(2+)-binding protein that upregulates synthesis of cGMP in photoreceptors. The known mammalian GCAPs are more than 90% similar, consisting of 201 to 205 amino acids, and containing 3 identically conserved Ca(2+)-binding sites. The GUCA1A gene, also termed GCAP1, is transcribed into a single 1.7-kb mRNA species detectable only in the retina. In a 4-generation British family with typical clinical features of autosomal domit cone dystrophy a tyr99-to-cys mutation) in the GUCA1A gene has been identified. Another family with a pro50-to-leu mutation in GUCA1A demonstrated phenotypic variability ranging from mild photophobia to rod-cone dystrophy. The mutant protein could activate guanylate cyclase 1 (GUCY2D) and displayed similar calcium sensitivity to wildtype protein. However, there was a marked increase in the susceptibility to protease degradation and a reduction in the thermal stability of the pro50-to-leu mutation, which may depress cellular concentration and thereby contribute to retinal cell mortality.Specifications
| GCAP1 | |
| Polyclonal | |
| Unconjugated | |
| GUCA1A | |
| C6orf131, COD3, GCAP, GCAP 1, GCAP1, Guanylate cyclase activator 1A, GUCA, GUCA1, GUCA1A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14913, 2978, 301233 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43080, P43081 | |
| Guca1a | |
| GCAP1 Fusion Protein Ag4017 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction