missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCAP1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00054C
This item is not returnable.
View return policy
Description
GCAP1 Polyclonal specifically detects GCAP1 in Human samples. It is validated for ELISA.
Specifications
| GCAP1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 650 | |
| C6orf131, chromosome 6 open reading frame 131, CORD14, GCAP 1, GCAP1photoreceptor 1, GCAPGUCA, Guanylate cyclase activator 1A, guanylate cyclase activator 1A (retina), guanylin 1, retina, guanylyl cyclase-activating protein 1, GUCA1 | |
| Produced in rabbits immunized with purified, recombinant GCAP1 (P43080; Met1-Gly201). | |
| 0.1 mL | |
| Vision | |
| 2978 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction