missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519802
This item is not returnable.
View return policy
Description
This antibody is predicted to react with pig based on sequence homology.
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
Specifications
| GC | |
| Polyclonal | |
| Unconjugated | |
| GC | |
| DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2638 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| P02774 | |
| GC | |
| Synthetic peptide conjugated to KLH derived from within residues 450 to the C-terminus of Human Vitamin D Binding Protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction