missing translation for 'onlineSavingsMsg'
Learn More

SGE™ GC Inlet Liner Accessories

Product Code. 10085360
Change view
Click to view available options
For Use With (Equipment):
All Liners Except SG092004, SG092235 Agilent, Thermo Trace 1300
Perkin-Elmer, Suitable for 6.2 mm OD Inlet Liners
Varian 1078, 1079 Injectors
For Use With (Application):
Graphite Sealing Ring
Viton O-Ring
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) For Use With (Application) unitSize
10085360 All Liners Except SG092004, SG092235 Agilent, Thermo Trace 1300 Graphite Sealing Ring Each
10113022 Varian 1078, 1079 Injectors Graphite Sealing Ring Pack of 10
12945711 Perkin-Elmer, Suitable for 6.2 mm OD Inlet Liners Viton O-Ring Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10085360 Supplier SGE™ Supplier No. 0726005
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories for use with GC inlet liners

  • Viton™ O-Ring—Can be used up to temperatures of 300°C (572°F)
  • Graphite Sealing Rings—Can be used up to temperatures of 450°C (842°F)

Specifications

Type SGE GC Inlet Liner Accessory
For Use With (Application) Graphite Sealing Ring
Temperature (Metric) Max 450°C
For Use With (Equipment) All Liners Except SG092004, SG092235 Agilent, Thermo Trace 1300
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.