missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GC/GC-MS Start-up Kit

Product Code. 31903478 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
TRACE 1300/1600 Series GC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31903478 TRACE 1300/1600 Series GC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31903478 Supplier Thermo Scientific™ Supplier No. 19050720
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Start-up kit for TRACE 1300/1600 GC/GC-MS systems including fittings, tubing, and tools required for column connection and initial installation.

  • Designed for TRACE 1300 and TRACE 1600 GC/GC-MS systems
  • Provides essential components for system start-up and column connection
  • Includes brass front and back ferrules (1/8 in. OD) for secure, leak-free connections
  • Includes retaining nuts for 1/8 in. OD metal columns
  • Includes brass union for 1/8 in. tubing connections
  • Includes plugs for 1/8 in. tube fittings
  • Supplied with pre-cleaned 1/8 in. OD tubing for installation
  • Includes installation tools: 7/16 in. wrench and tube cutter (1/8 in. - 5/8 in.)
  • Supports reliable system setup and ensures proper gas flow integrity
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) TRACE 1300/1600 Series GC
Includes Tubing, Ferrules, Plugs, Installation Tools
Sterility Non-sterile
Color Light Brown
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.