Learn More
Trajan Scientific and Medical™ GC ferrule OGF16-16 one piece graphite 1/16 inch x 1/16 inch fitting size x hole size SGE
Product Code. 10656401
Change view
Click to view available options
For Use With (Equipment):
1/16 in. Fittings
Unit Size:
Pack of 10
Product Code. For Use With (Equipment) unitSize
10656401 1/16 in. Fittings Pack of 10
1 options
Product Code. 10656401 Supplier Trajan Scientific and Medical™ Supplier No. 072623
Only null left
Add to basket
Add to basket
Provide leak-free system connections. Trajan Scientific and Medical™ Straight Ferrule for 1/16 in. Fittings lead to improved chromatography for non-MS systems.
  • Easy to use
  • Forms reliable seal
  • Reusable
  • Soft material, forms soft grip to the capillary column
  • Low emissions
Specifications
Type GC Ferrule
Description 1/16 in. I.D. hole
Diameter (English) Inside 1/16 in.
Length (Metric) 3.7 mm
Width (Metric) 3.2 mm
For Use With (Equipment) 1/16 in. Fittings
Material Graphite
Quantity 10/Pk.
Diameter (Metric) Inside 1.59 mm