missing translation for 'onlineSavingsMsg'
Learn More

SGE Analytical Science™ GC Deactivated Inlet Liner

Product Code. 12907141
Change view
Click to view available options
Diameter (Metric) Inner:
4.0 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
12907141 4.0 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12907141 Supplier SGE Analytical Science™ Supplier No. 09200201
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use these liners for a variety of applications. SGE Analytical Science™ GC Deactivated Inlet Liners are tested for activity using the EPA 8081B method, ensuring each batch of inlet liners has less than 3% Endrin breakdown.

  • GC Deactivated Inlet Liner for Agilent S/S
  • Deactivated with a high temperature (>400°C) gas phase reagent
  • Split/Splitless Focusliner
  • 4.0mm I.D.
  • Batch certificate included with every pack

Specifications

For Use With (Application) Agilent Instruments
Length (Metric) 78.5 mm
Product Type GC Deactivated Inlet Liner
Diameter (Metric) Inner 4.0 mm
Diameter (Metric) Outer 6.3 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.