Learn More
SGE Analytical Science™ GC Autosampler Syringe
Product Code. 10741232
Click to view available options
Volume (Metric):
0.5 μL
Unit Size:
Each
Product Code. 10741232
Brand: SGE Analytical Science™ 000478
Designed and tested to meet critical autosampler specifications. SGE Analytical Science™ GC Autosampler Syringe feature a unique plunger design which minimizes bending and seizing, and the cone tip needle point style has been developed specifically to withstand multiple fast septum injections.
  • AutoSystem™ and Clarus™ Removable Needle
  • 0.5μL syringe
  • FOR PE
  • 0.63mm O.D.
Specifications
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type Autosampler Syringe
Volume (Metric) 0.5 μL
Needle Point Style Cone