missing translation for 'onlineSavingsMsg'
Learn More

GBR 12935 Dihydrochloride, TRC

Product Code. 30402816
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402816 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402816 Supplier Toronto Research Chemicals Supplier No. TRCG26800325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[2-(Diphenylmethoxy)ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride
CAS 67469-81-2
Molecular Formula C28H34N2O . 2HCl
Quantity 25 mg
Synonym GBR 12935 Dihydrochloride, 1-[2-(Diphenylmethoxy)ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride
InChI Formula InChI=1S/C28H34N2O.ClH/c1-4-11-25(12-5-1)13-10-18-29-19-21-30(22-20-29)23-24-31-28(26-14-6-2-7-15-26)27-16-8-3-9-17-27;/h1-9,11-12,14-17,28H,10,13,18-24H2;1H
SMILES Cl.C(CN1CCN(CCOC(c2ccccc2)c3ccccc3)CC1)Cc4ccccc4
IUPAC Name 1-(2-benzhydryloxyethyl)-4-(3-phenylpropyl)piperazine;hydrochloride
Molecular Weight (g/mol) 487.5
Formula Weight 450.2438
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.