missing translation for 'onlineSavingsMsg'
Learn More

GBR-12909 Dihydrochloride, TRC

Product Code. 17971215
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971215 50 mg 50mg
17943795 5 mg 5mg
17912065 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971215 Supplier Toronto Research Chemicals Supplier No. TRCG26800550MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67469-78-7
Molecular Formula C28 H32 F2 N2 O . 2(H Cl)
Molecular Weight (g/mol) 523.48
Synonym GBR-12909 Dihydrochloride, 1-[2-[Bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride (9CI), GBR 12909, I 893, Vanoxerine Dihydrochloride, 1-[2-[Bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)-piperazine Hydrochloride (1:2)
IUPAC Name 1-[2-[bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine;hydrochloride
SMILES Cl.Cl.Fc1ccc(cc1)C(OCCN2CCN(CCCc3ccccc3)CC2)c4ccc(F)cc4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H32F2N2O.2ClH/c29-26-12-8-24(9-13-26)28(25-10-14-27(30)15-11-25)33-22-21-32-19-17-31(18-20-32)16-4-7-23-5-2-1-3-6-23;;/h1-3,5-6,8-15,28H,4,7,16-22H2;2*1H
Formula Weight 522.2016
Chemical Name or Material 1-[2-[Bis(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)-piperazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.