missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GBR 12909 dihydrochloride
GREENER_CHOICE

Product Code. 18728896
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18728896 50 mg 50mg
18767696 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18728896 Supplier Tocris Bioscience™ Supplier No. 0421/50
  • ONLINE EXCLUSIVE PRICE
    £389.00
    £367.50 / 50mg
    Save £21.50
    5% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective DA uptake inhibitor; also σ ligand

GBR 12909 dihydrochloride is a potent, competitive inhibitor of dopamine uptake (Ki = 1 nM for inhibition of striatal dopamine uptake). Has >100-fold lower affinity for the noradrenaline and 5-HT uptake carriers. Also a potent sigma ligand (IC50 = 48 nM). Centrally active following systemic administration.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[2-[Bis-(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride
CAS 67469-78-7
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 50mM in DMSO
Molecular Formula C28H32F2N2O.2HCl
Formula Weight Observed MW: 523.49

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.