missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GBP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534541
This item is not returnable.
View return policy
Description
A suggested positive control is rat liver tissue lysate. PA5-34541 can be used with blocking peptide PEP-1584.
The guanylate-binding proteins (GBPs) are a family of interferon-induced GTP-binding proteins function in innate immunity against microbial and viral pathogens. In humans, there are seven GBPs from hGBP1 to hGBP7. Human GBP1 and GBP2 have been shown to exhibit antiviral activity as well as being able to regulate the inhibition of proliferation and invasion of endothelial cells in response to interferon (IFN). GBP5 promotes selective NALP3 inflammasome responses to pathogenic bacteria and soluble but not crystalline inflammasome priming agents.
Specifications
| GBP5 | |
| Polyclonal | |
| Unconjugated | |
| GBP5 | |
| 5330409J06Rik; GBP5; GBP-5; Gbp5a; GBP-TA antigen; GTP-binding protein 5; Guanine nucleotide-binding protein 5; guanylate binding protein 5; guanylate nucleotide binding protein 5; guanylate-binding protein 5; muGBP-5; UNQ2427/PRO4987 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115362, 229898, 362050 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8CFB4, Q96PP8 | |
| GBP5 | |
| A 16 amino acid peptide near the carboxy terminus of human GBP5. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction