Learn More
Invitrogen™ GBP4 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113284
Product Code. 17151144
  • £400.00 / 100µg
Description
GBP4 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA
Guanylate-binding proteins, such as GBP4, are induced by interferon and hydrolyze GTP to both GDP and GMP.[supplied by OMIM, Dec 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GBP4
Polyclonal
Unconjugated
GBP4
AW228052; Gbp3; GBP4; GBP-4; GTP-binding protein 4; guanine nucleotide-binding protein 4; guanylate binding protein 4; guanylate nucleotide binding protein 3; guanylate nucleotide binding protein 4; guanylate-binding protein 4; guanylate-binding protein 4 homolog; macrophage activation 2; Mag-2; mKIAA4245; Mpa2; Mpa-2
Rabbit
Protein G
RUO
115361
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q96PP9
GBP4
Recombinant Human Guanylate-binding protein 4 protein (494-603AA).