missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP4 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00115361-B01P
This item is not returnable.
View return policy
Description
GBP4 Polyclonal antibody specifically detects GBP4 in Human samples. It is validated for Western Blot
Specifications
| GBP4 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH70055.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 115361 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| GBP-4, GTP-binding protein 4, Guanine nucleotide-binding protein 4, guanylate binding protein 4, guanylate-binding protein 4, Mpa2 | |
| GBP4 (AAH70055.1, 1 a.a. - 640 a.a.) full-length human protein. MGERTLHAAVPTPGYPESESIMMAPICLVENQEEQLTVNSKALEILDKISQPVVVVAIVGLYRTGKSYLMNRLAGKRNGFPLGSTVQSETKGIWMWCVPHLSKPNHTLVLLDTEGLGDVEKSNPKNDSWIFALAVLLSSSFVYNSVSTINHQALEQLHYVTELAELIRAKSCPRPDEAEDSSEFASFFPDFIWTVRDFTLELKLDGNPITEDEYLENALKLIPGKNPKIQNSNMPRECIRHFFRKRKCFVFDRPTNDKQYLNHMDEVPEENLERHFLMQSDNFCSYIFTHAKTKTLRKGIIVTGKRLGTLVVTYVDAINSGAVPCLENAVTALAQLENPAAVQRAADHYSQQMAQQLRLPTDTLQELLDVHAACEREAIAVFMEHSFKDENHEFQKKLVDTIEKKKGDFVLQNEEASAKYCQAELKRLSEHLTESILRGIFSVPGGHNLYLEEKKQVEWDYKLVPRKGVKANEVLQNFLQSQVVVEESILQSDKALTAGEKSIAAERAMKEAAEKEQELLREKQKEQQQMMEAQERSFQEYMAQMEKKLEEERENLLREHERLLKHKLKVQEEMLKEEFQKKSEQLNKEINQLKEKIESTKNEQLRLLKILDMASNIMIVTLPGASKLLGVGTKYLGSRI | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction