missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522238
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLa, HepG2, Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the guanylate-binding protein (GBP) family. GBPs specifically bind guanine nucleotides (GMP, GDP, and GTP) and contain two of the three consensus motifs found in typical GTP-binding proteins. The encoded protein interacts with a member of the germinal center kinase family. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| GBP3 | |
| Polyclonal | |
| Unconjugated | |
| GBP3 | |
| GBP3; GBP-3; GTP-binding protein 3; guanine nucleotide-binding protein 3; guanylate binding protein 3; guanylate-binding protein 3; guanylate-binding protein 3 delta C; RP4-531M19.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.81 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H0R5 | |
| GBP3 | |
| Recombinant protein encompassing a sequence within the center region of human GBP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction