missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11854-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.Specifications
| GBP2 | |
| Polyclonal | |
| Unconjugated | |
| GBP2 | |
| GBP 2, GBP2, GTP binding protein 2, HuGBP 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171164, 2634 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P32456, Q63663 | |
| Gbp2 | |
| GBP2 Fusion Protein Ag2425 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction