missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112426
This item is not returnable.
View return policy
Description
GBP2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
Specifications
| GBP2 | |
| Polyclonal | |
| Unconjugated | |
| GBP2 | |
| GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2634 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P32456 | |
| GBP2 | |
| Recombinant Human Guanylate-binding protein 2 protein (100-259AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction