missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00230-50ul
This item is not returnable.
View return policy
Description
GBP2 Polyclonal antibody specifically detects GBP2 in Human samples. It is validated for ELISA
Specifications
| GBP2 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| GBP-2, GTP-binding protein 2, Guanine nucleotide-binding protein 2, guanylate binding protein 2, interferon-inducible, huGBP-2, interferon-induced guanylate-binding protein 2 | |
| Produced in rabbits immunized with purified, recombinant Human GBP2 (Accession#: AAH73163.1; Met1-Cys588) | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 2634 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction