missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528307
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GBP1 or guanlyate binding protein 1 is a 68 kDa GTPase and member of the GTPase1 family of molecules. GBP1 expression is induced by Interferon g as well as other inflammatory cytokines including IL-1a, IL-1b and TNF-a. Induction of GBP1 expression by poly I:C, a prototypical TLR3 ligand, as well as GBP1 expression dependency on NF-kB and IFNa response elements implicate TLR pathway involvement. Inversely proportional expression of GBP1 and MMP-1 as well as other related molecules to adhesion and migration show a clear functional role of GBP1 in suppressing angiogenesis and migration especially of endothelial cells. As a regulatory molecule it is centrally involved in signal transduction of inflammatory and cell migration networks.
Specifications
| GBP1 | |
| Polyclonal | |
| Unconjugated | |
| GBP1 | |
| GBP1; Gbp-1; Gbp2b; GTP-binding protein 1; guanine nucleotide-binding protein 1; guanylate binding protein 1; guanylate binding protein 1, interferon-inducible; guanylate binding protein 1, interferon-inducible, 67kDa; guanylate binding protein 2b; guanylate nucleotide binding protein 1; guanylate-binding protein 1; huGBP-1; interferon-gamma-inducible protein MAG-1; interferon-induced guanylate-binding protein 1; Mag-1; Mpa1; Mpa-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2633 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P32455 | |
| GBP1 | |
| Recombinant protein encompassing a sequence within the center region of human GBP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction