missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBP1 Antibody (SR2121), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22422-100ul
This item is not returnable.
View return policy
Description
GBP1 Monoclonal antibody specifically detects GBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| GBP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR2121 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| GBP-1, GTP-binding protein 1, Guanine nucleotide-binding protein 1, guanylate binding protein 1, interferon-inducible, guanylate binding protein 1, interferon-inducible, 67kDa, huGBP-1, interferon-induced guanylate-binding protein 1 | |
| A synthesized peptide derived from human GBP1 (Uniprot #: P32455) | |
| 100 μg | |
| Immunology, Innate Immunity, Signal Transduction | |
| 2633 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction