missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GBP1 Antibody (GBP1/7618), Novus Biologicals™
Mouse Monoclonal Antibody
£178.00 - £351.00
Specifications
| Antigen | GBP1 |
|---|---|
| Clone | GBP1/7618 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043608
|
Novus Biologicals™
NBP3-26793-100ug |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043341
|
Novus Biologicals™
NBP3-26793-20ug |
20 μg |
£178.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GBP1 Monoclonal antibody specifically detects GBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GBP1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 2633 | |
| IgG | |
| Protein A or G purified |
| GBP1/7618 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GBP-1, GTP-binding protein 1, Guanine nucleotide-binding protein 1, guanylate binding protein 1, interferon-inducible, guanylate binding protein 1, interferon-inducible, 67kDa, huGBP-1, interferon-induced guanylate-binding protein 1 | |
| Recombinant fragment (around aa400-592) of human GBP1 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title