missing translation for 'onlineSavingsMsg'
Learn More

GBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18317072 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18317072 20 μg 20µL
18357761 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18317072 Supplier Novus Biologicals Supplier No. NBP30345020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GBL Polyclonal antibody specifically detects GBL in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBL
Applications Western Blot, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Knockout Validated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-326 of human GBL (NP_001186103.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 64223
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.