Learn More
GBL Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98484PE
This item is not returnable.
View return policy
Description
GBL Polyclonal antibody specifically detects GBL in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Store at 4C in the dark. |