missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBL Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2124R
This item is not returnable.
View return policy
Description
GBL Polyclonal antibody specifically detects GBL in Human samples. It is validated for Western Blot
Specifications
| GBL | |
| Polyclonal | |
| Western Blot | |
| G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1 | |
| The immunogen recognized by this antibody maps to a region between residues 275 and the C-terminus (residue 326) of human G protein Beta subunit-like using the numbering given in entry NP_071767.2 (GeneID 64223). | |
| 0.1 mL | |
| Cancer, Hypoxia, mTOR Pathway | |
| 64223 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction