missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GBAS Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858802
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858802 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858802 Supplier Invitrogen™ Supplier No. PA520656

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human skeletal muscle tissue lysate. PA5-20656 can be used with blocking peptide PEP-0776.

GBAS encodes a protein belonging to the NipSnap family, which is believed to participate in vesicular transport. This protein is primarily found in mitochondria and is involved in oxidative phosphorylation. A non-functional version of this gene is located on the long arm of chromosome 2. Multiple isoforms of this gene have been identified due to alternative splicing. NIPSNAP2 is linked to several diseases such as Phosphoserine Phosphatase Deficiency and Mast Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBAS
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NIPSNAP2
Gene Accession No. O55126, O75323
Gene Alias 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; AV006093; GBAS; glioblastoma amplified sequence; glioblastoma-amplified sequence; nipsnap homolog 2; NIPSNAP2; Protein NipSnap homolog 2
Gene Symbols NIPSNAP2
Host Species Rabbit
Immunogen A 14 amino acid peptide near the center of human NIPSNAP2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14467, 2631, 498174
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.