missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GBA Recombinant Rabbit Monoclonal Antibody (23GB5775), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GBA |
|---|---|
| Clone | 23GB5775 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089540
![]() |
Invitrogen™
MA552914 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088875
|
Invitrogen™
MA552915 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GBA Recombinant Monoclonal antibody specifically detects GBA in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GBA | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P04062, P17439 | |
| 14466, 2629, 684536 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5775 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| acid beta glucosidase; acid beta-glucosidase; Alglucerase; betaGC; beta-GC; Beta-glucocerebrosidase; Cholesterol glucosyltransferase; Cholesteryl-beta-glucosidase; D-glucosyl-N-acylsphingosine glucohydrolase; Gba; GBA1; GBAP; GBAP1; GC; GCase; GCB; GLUC; glucocerebrosidase; glucosidase, beta, acid; glucosylceramidase; glucosylceramidase beta; glucosylceramidase beta pseudogene 1; glucosylceramidase-like protein; imiglucerase; Lysosomal acid GCase; Lysosomal acid glucosylceramidase; lysosomal glucocerebrosidase; SGTase | |
| A synthesized peptide derived from human GBA (497-536AA). | |
| Primary | |
| -20°C | |
| GBA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title