missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | GBA |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16810185
|
Proteintech
20622-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16800185
|
Proteintech
20622-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GBA encodes a lysosomal membrane protein that cleaves the beta-glucosidic linkage of glycosylceramide, an intermediate in glycolipid metabolism. Mutations in this gene cause Gaucher disease, a lysosomal storage disease characterized by an accumulation of glucocerebrosides. A related pseudogene is approximately 12 kb downstream of this gene on chromosome 1. Alternative splicing results in multiple transcript variants.Specifications
| GBA | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| GBAP | |
| GBAP1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 2630 | |
| GBA Fusion Protein Ag13981 | |
| Primary | |
| -20°C | |
| GBAP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title