missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBA Polyclonal Antibody

GBA Polyclonal Antibody
Brand: Invitrogen PA572598
This item is not returnable.
View return policy
Description
GBA Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| GBA | |
| Polyclonal | |
| Unconjugated | |
| GBA | |
| acid beta glucosidase; acid beta-glucosidase; Alglucerase; betaGC; beta-GC; Beta-glucocerebrosidase; Cholesterol glucosyltransferase; Cholesteryl-beta-glucosidase; D-glucosyl-N-acylsphingosine glucohydrolase; Gba; GBA1; GBAP; GBAP1; GC; GCase; GCB; GLUC; glucocerebrosidase; glucosidase, beta, acid; glucosylceramidase; glucosylceramidase beta; glucosylceramidase beta pseudogene 1; glucosylceramidase-like protein; imiglucerase; Lysosomal acid GCase; Lysosomal acid glucosylceramidase; lysosomal glucocerebrosidase; SGTase | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 14466, 2629 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P04062, P17439 | |
| GBA | |
| KLH conjugated synthetic peptide between 337-365 amino acids of human GC. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction