missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB/T21311-2007 Nitrofuran Metabolites Labeled Mixture 346 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 1-Amino Hydantoin-13C3∣3-Amino-2-oxazolidinone-d4∣3-Amino-5-morpholinomethyl-2-oxazolidinone D5 (AMOZ D5)∣Semicarbazide hydrochloride-13C,15N2 |
| CAS | 957509-31-8, 1188331-23-8, 1017793-94-0, 1173020-16-0 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1-Amino Hydantoin-13C3 100 μg/mL∣3-Amino-2-oxazolidinone-d4 100 μg/mL∣3-Amino-5-morpholinomethyl-2-oxazolidinone D5 (AMOZ D5) 100 μg/mL∣Semicarbazide hydrochloride-13C,15N2 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?