missing translation for 'onlineSavingsMsg'
Learn More

GB/T 5750.9-2006 Pyrethroids Pesticide Mixture 557 50 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30324962
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324962 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324962 Supplier Dr. Ehrenstorfer Supplier No. DREA50000557AL
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Fenpropathrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Permethrin
CAS 39515-41-8, 52315-07-8, 52918-63-5, 51630-58-1, 91465-08-6, 52645-53-1
Analyte or Component Names (±)-Fenpropathrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Permethrin
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (±)-Fenpropathrin 50 μg/mL∣Cypermethrin 50 μg/mL∣Deltamethrin 50 μg/mL∣Fenvalerate 50 μg/mL∣lambda-Cyhalothrin 50 μg/mL∣Permethrin 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.