missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GB/T 20759-2006 Sulfonamides Mixture 583 100 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Sulfacetamide∣Sulfachloropyridazine∣Sulfadiazine∣Sulfadimethoxine∣Sulfadimidine∣Sulfadoxine∣Sulfamerazine∣Sulfameter∣Sulfamethizole∣Sulfamethoxazole∣Sulfamethoxypyridazine∣Sulfamonomethoxine∣Sulfaphenazole∣Sulfapyridine∣Sulfathiazole∣Sulfisoxazole |
| CAS | 144-80-9, 80-32-0, 68-35-9, 122-11-2, 57-68-1, 2447-57-6 |
| Analyte or Component Names | Sulfacetamide∣Sulfachloropyridazine∣Sulfadiazine∣Sulfadimethoxine∣Sulfadimidine∣Sulfadoxine∣Sulfamerazine∣Sulfameter∣Sulfamethizole∣Sulfamethoxazole∣Sulfamethoxypyridazine∣Sulfamonomethoxine∣Sulfaphenazole∣Sulfapyridine∣Sulfathiazole∣Sulfisoxazole |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Sulfacetamide 100 μg/mL∣Sulfachloropyridazine 100 μg/mL∣Sulfadiazine 100 μg/mL∣Sulfadimethoxine 100 μg/mL∣Sulfadimidine 100 μg/mL∣Sulfadoxine 100 μg/mL∣Sulfamerazine 100 μg/mL∣Sulfameter 100 μg/mL∣Sulfamethizole 100 μg/mL∣Sulfamethoxazole 100 μg/mL∣Sulfamethoxypyridazine 100 μg/mL∣Sulfamonomethoxine 100 μg/mL∣Sulfaphenazole 100 μg/mL∣Sulfapyridine 100 μg/mL∣Sulfathiazole 100 μg/mL∣Sulfisoxazole 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction