missing translation for 'onlineSavingsMsg'
Learn More

GB/T 14848-2017 Pesticides Mixture 522 100 μg/mL in Toluene, Dr. Ehrenstorfer

Product Code. 30316847
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316847 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316847 Supplier Dr. Ehrenstorfer Supplier No. DREA50000522TO
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorophenoxyacetic acid∣Aldicarb∣Atrazine∣Carbofuran∣Chlorothalonil∣Chlorpyrifos∣Dichlorvos∣Dimethoate∣Heptachlor∣Malathion∣Parathion-methyl
CAS 94-75-7, 116-06-3, 1912-24-9, 1563-66-2, 1897-45-6, 2921-88-2
Analyte or Component Names 2,4-Dichlorophenoxyacetic acid∣Aldicarb∣Atrazine∣Carbofuran∣Chlorothalonil∣Chlorpyrifos∣Dichlorvos∣Dimethoate∣Heptachlor∣Malathion∣Parathion-methyl
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4-Dichlorophenoxyacetic acid 100 μg/mL∣Aldicarb 100 μg/mL∣Atrazine 100 μg/mL∣Carbofuran 100 μg/mL∣Chlorothalonil 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Dichlorvos 100 μg/mL∣Dimethoate 100 μg/mL∣Heptachlor 100 μg/mL∣Malathion 100 μg/mL∣Parathion-methyl 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.