missing translation for 'onlineSavingsMsg'
Learn More

GB 31660.2-2019 Antibiotics Mixture 168 30-100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30324623
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30324623

Brand: Dr. Ehrenstorfer DREA50000168ME

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-tert-Octylphenol∣Bisphenol A∣Diethylstilbestrol∣Estradiol∣Estriol∣Estrone∣Ethinylestradiol∣n-Nonylphenol
CAS 140-66-9, 80-05-7, 56-53-1, 50-28-2, 50-27-1, 53-16-7
Analyte or Component Names 4-tert-Octylphenol∣Bisphenol A∣Diethylstilbestrol∣Estradiol∣Estriol∣Estrone∣Ethinylestradiol∣n-Nonylphenol
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Bisphenol A 30 μg/mL∣n-Nonylphenol 30 μg/mL∣4-tert-Octylphenol 50 μg/mL∣Diethylstilbestrol 50 μg/mL∣Estradiol 100 μg/mL∣Estriol 100 μg/mL∣Estrone 100 μg/mL∣Ethinylestradiol 100 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.